Thermo Scientific™

Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform, Pd 20.6%, Thermo Scientific Chemicals

Catalog number: 044969.06
5 g, Each
Thermo Scientific™

Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform, Pd 20.6%, Thermo Scientific Chemicals

Catalog number: 044969.06
5 g, Each
Quantity
Catalog number: 044969.06
also known as 044969-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
52522-40-4
IUPAC Name
tris(1,5-diphenylpenta-1,4-dien-3-one) trichloromethane dipalladium
Molecular Formula
C52H43Cl3O3Pd2
InChI Key
LNAMMBFJMYMQTO-UHFFFAOYSA-N
SMILES
[Pd].[Pd].ClC(Cl)Cl.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1
Assay (unspecified)
Pd 20.6% min

Description

Tris(dibenzylideneacetone)dipalladium(0) is used in Synthesis of nanosized palladium phosphides upon interaction with white phosphorous, Preparation of palladium triphenylphosphine carbonyl cluster complexes, Precursor for synthesis of functionalized multiwalled carbon nanotube-palladium complexes used as catalysts for Heck coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text