Thermo Scientific Chemicals

Dichlorobis(tri-o-tolylphosphine)palladium(II), 95%, Thermo Scientific Chemicals

Catalog number: 044976.03
1 g, Each
Thermo Scientific Chemicals

Dichlorobis(tri-o-tolylphosphine)palladium(II), 95%, Thermo Scientific Chemicals

Catalog number: 044976.03
1 g, Each
Quantity
Catalog number: 044976.03
also known as 044976-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
40691-33-6
IUPAC Name
dichloropalladiumbis(ylium); bis(tris(2-methylphenyl)phosphane)
Molecular Formula
C42H42Cl2P2Pd
InChI Key
GMLMZLGOMMIIMA-UHFFFAOYSA-L
SMILES
Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1
Form
Powder
Assay (special)
≥94%

Description

Dichlorobis(tri-o-tolylphosphine)palladium(II) is used in reaction of tributyltin enolates, prepared in situ from tributyltin methoxide and enol acetates, with aryl bromides, coupling reaction of aryl bromides with vinylic acetates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text