Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II), Pd 13% min, Thermo Scientific Chemicals
Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II), Pd 13% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II), Pd 13% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 044977.03
also known as 044977-03
Price (USD)/ Each
128.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
128.00
Add to cart
Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II), Pd 13% min, Thermo Scientific Chemicals
Catalog number044977.03
Price (USD)/ Each
128.00
-
Add to cart
Chemical Identifiers
CAS205319-06-8
IUPAC Namedichloropalladiumbis(ylium); {2-[2-(diphenylphosphanyl)phenoxy]phenyl}diphenylphosphane
Molecular FormulaC36H28Cl2OP2Pd
InChI KeyQZIOEBWDZNUTLZ-UHFFFAOYSA-L
SMILESCl[Pd++]Cl.O(C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)3.40-4.40% Hydrogen
Assay (unspecified)59.40-61.40% Carbon
Assay (unspecified)8.90-10.90% Chlorine
Assay (unspecified)Pd 13.80-15.80%
Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II) is used as a catalyst for cross-coupling reaction of 2-brom-1,3-dienes to prepare high stereoselectivity of conjugated Z,E dienes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only