Thermo Scientific Chemicals

Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II), Pd 13% min, Thermo Scientific Chemicals

Catalog number: 044977.06
5 g, Each
Thermo Scientific Chemicals

Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II), Pd 13% min, Thermo Scientific Chemicals

Catalog number: 044977.06
5 g, Each
Quantity
Catalog number: 044977.06
also known as 044977-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
205319-06-8
IUPAC Name
dichloropalladiumbis(ylium); {2-[2-(diphenylphosphanyl)phenoxy]phenyl}diphenylphosphane
Molecular Formula
C36H28Cl2OP2Pd
InChI Key
QZIOEBWDZNUTLZ-UHFFFAOYSA-L
SMILES
Cl[Pd++]Cl.O(C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Assay (unspecified)
3.40-4.40% Hydrogen
Assay (unspecified)
59.40-61.40% Carbon
Assay (unspecified)
8.90-10.90% Chlorine
Assay (unspecified)
Pd 13.80-15.80%

Description

Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II) is used as a catalyst for cross-coupling reaction of 2-brom-1,3-dienes to prepare high stereoselectivity of conjugated Z,E dienes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text