Aluminum shot, nominally 9.5mm (0.375in) flattened/spherical shape, 99.99% (metals basis), Thermo Scientific Chemicals
Aluminum shot, nominally 9.5mm (0.375in) flattened/spherical shape, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum shot, nominally 9.5mm (0.375in) flattened/spherical shape, 99.99% (metals basis), Thermo Scientific Chemicals

Spheroid shape pellet 3/8 in. ±1/4 in.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Catalog number 045001.36
also known as 045001-36
Price (USD)/ Each
84.65
Online exclusive
94.30 
Save 9.65 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
84.65
Online exclusive
94.30 
Save 9.65 (10%)
Add to cart
Aluminum shot, nominally 9.5mm (0.375in) flattened/spherical shape, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number045001.36
Price (USD)/ Each
84.65
Online exclusive
94.30 
Save 9.65 (10%)
-
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
FormSpheroid shape pellet, nominally 9.5 mm
Total Metal Impurities≤0.01%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only