Thermo Scientific Chemicals
Sapphire substrate, 50.8mm dia x 0.432mm thick, EPI polished one side, C-axis, HEMCOR single crystal
Catalog number: 045019.KT
1 Ea., Each
Thermo Scientific Chemicals
Sapphire substrate, 50.8mm dia x 0.432mm thick, EPI polished one side, C-axis, HEMCOR single crystal
Catalog number: 045019.KT
1 Ea., Each
Quantity
Catalog number: 045019.KT
also known as 045019-KT
Price (EUR)
Quantity
-
Comment
50.8mm dia x 0.432mm thick
Comment
C-axis
Description
Sapphire is used as an insulating substrate upon which to deposit silicon to make the integrated circuits, high-power radio-frequency (RF) applications such as those found in cellular telephones, public-safety band radios, and satellite communication systems, used in the semiconductor industry, blue light-emitting diodes.