Thermo Scientific Chemicals

Gallium(III) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 045026.06
5 g, Each
Thermo Scientific Chemicals

Gallium(III) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 045026.06
5 g, Each
Quantity
Catalog number: 045026.06
also known as 045026-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13450-90-3
IUPAC Name
gallium(3+) trichloride
Molecular Formula
Cl3Ga
InChI Key
UPWPDUACHOATKO-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Ga+3]
Appearance (Color)
White
Form
Granules
Residual water
< 100 ppm
Total Metal Impurities
0.001% max.

Description

Gallium(III) chloride is a catalyst for a number of transformations including the ring opening and recyclization of epoxides and the trimerization of alkynes. It is used to detect the solar neutrions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text