Antimony broken rod, 12.7mm (0.50in) dia x various lengths, 99.8% (metals basis), Thermo Scientific Chemicals
Antimony broken rod, 12.7mm (0.50in) dia x various lengths, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Antimony broken rod, 12.7mm (0.50in) dia x various lengths, 99.8% (metals basis), Thermo Scientific Chemicals

Form: Broken pieces from cast rod
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
Catalog number 045047.A3
also known as 045047-A3
Price (USD)/ Each
618.65
Online exclusive
687.00 
Save 68.35 (10%)
-
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (USD)/ Each
618.65
Online exclusive
687.00 
Save 68.35 (10%)
Add to cart
Antimony broken rod, 12.7mm (0.50in) dia x various lengths, 99.8% (metals basis), Thermo Scientific Chemicals
Catalog number045047.A3
Price (USD)/ Each
618.65
Online exclusive
687.00 
Save 68.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-36-0
IUPAC Nameantimony
Molecular FormulaSb
InChI KeyWATWJIUSRGPENY-UHFFFAOYSA-N
SMILES[Sb]
View more
SpecificationsSpecification SheetSpecification Sheet
FormBroken pieces from cast rod, 0.5in diameter and various lengths
Total Metal Impurities≤0.2%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only