Thermo Scientific Chemicals

Poly(styrene sulfonic acid) sodium salt, M.W. 500,000, Thermo Scientific Chemicals

Catalog number: 045050.22
100 g, Each
Thermo Scientific Chemicals

Poly(styrene sulfonic acid) sodium salt, M.W. 500,000, Thermo Scientific Chemicals

Catalog number: 045050.22
100 g, Each
Quantity
Catalog number: 045050.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25704-18-1
Molecular Formula
(C8H7NaO3S)n
SMILES
[Na]OS(=O)(=O)C1=CC=C(C=C1)C(-*)C-*
MDL Number
MFCD00084449
Appearance (Color)
White to cream
Comment
Molecular Weight: 500,000 (approx.)
Infrared spectrum
Conforms
Volatiles
≤10.00%
Form
Powder

Description

Poly(styrene sulfonic acid) sodium salt is used in superplastifier in cement, ion exchange resins, membranes in fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text