Zirconium wire, 0.5mm (0.020in) dia, annealed, 99.2% (metals basis excluding Hf) Hf 4.5% max, Thermo Scientific Chemicals
Zirconium wire, 0.5mm (0.020in) dia, annealed, 99.2% (metals basis excluding Hf) Hf 4.5% max, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium wire, 0.5mm (0.020in) dia, annealed, 99.2% (metals basis excluding Hf) Hf 4.5% max, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
1 m
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045056.G5
Price (USD)/ Each
-
Quantity:
5 m
Request bulk or custom format
Chemical Identifiers
CAS7440-67-7
IUPAC Namezirconium
Molecular FormulaZr
InChI KeyQCWXUUIWCKQGHC-UHFFFAOYSA-N
SMILES[Zr]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace Metal: 99.2% (metals basis excluding Hf), Hf 4.5% max
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only