Thermo Scientific Chemicals

Arsenic(III) iodide, 98%, Thermo Scientific Chemicals

Catalog number: 045067.09
10 g, Each
Thermo Scientific Chemicals

Arsenic(III) iodide, 98%, Thermo Scientific Chemicals

Catalog number: 045067.09
10 g, Each
Quantity
Catalog number: 045067.09
also known as 045067-09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7784-45-4
IUPAC Name
triiodoarsane
Molecular Formula
AsI3
InChI Key
IKIBSPLDJGAHPX-UHFFFAOYSA-N
SMILES
I[As](I)I
Assay (unspecified)
97.6% min

Description

Arsenic(III) iodide is mainly used to treat rheumatism, arthritis, malaria, trypanosome infections, tuberculosis, and diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text