Lead granules, +30 mesh, 99.99% (metals basis), Thermo Scientific Chemicals
Lead granules, +30 mesh, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead granules, +30 mesh, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
Catalog number 045081.36
also known as 045081-36
Price (USD)/ Each
54.65
Online exclusive
60.00 
Save 5.35 (9%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
54.65
Online exclusive
60.00 
Save 5.35 (9%)
Add to cart
Lead granules, +30 mesh, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number045081.36
Price (USD)/ Each
54.65
Online exclusive
60.00 
Save 5.35 (9%)
-
Add to cart
Chemical Identifiers
CAS7439-92-1
IUPAC Namelead
Molecular FormulaPb
InChI KeyWABPQHHGFIMREM-UHFFFAOYSA-N
SMILES[Pb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only