Barium titanium oxide, 99.95% (metals basis), Thermo Scientific Chemicals
Barium titanium oxide, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium titanium oxide, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 045083.14
also known as 045083-14
Price (USD)/ Each
350.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
350.00
Add to cart
Barium titanium oxide, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number045083.14
Price (USD)/ Each
350.00
-
Add to cart
Chemical Identifiers
CAS12047-27-7
IUPAC Namebarium(2+) oxotitaniumbis(olate)
Molecular FormulaBaO3Ti
InChI KeyWNKMTAQXMLAYHX-UHFFFAOYSA-N
SMILES[Ba++].[O-][Ti]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only