Nickel Iron Molybdenum foil, 0.102mm (0.004in) thick, annealed
Nickel Iron Molybdenum foil, 0.102mm (0.004in) thick, annealed
Thermo Scientific Chemicals

Nickel Iron Molybdenum foil, 0.102mm (0.004in) thick, annealed

Form: approx. 40.1g/150 x 300mm | Ni:Fe:Mo
Have Questions?
Change viewbuttonViewtableView
Quantity:
150 x 300 mm
300 x 300 mm
300 x 600 mm
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045092.ATO
also known as 045092-@O
Price (USD)
-
Quantity:
300 x 600 mm
Request bulk or custom format
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeNi:Fe:Mo; 80:15:4.2 wt%
Chemical Name or MaterialNickel Iron Molybdenum foil
Health Hazard 1H317-H350-H372
Health Hazard 2GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
View more
It is most commonly used as a high temperature, corrosion resistant metal alloy in steel, cast iron and super alloys for the military and defense industry, semiconductor industry and specialty machine shops.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection