Nickel Iron Molybdenum foil, 0.102mm (0.004in) thick, annealed, Thermo Scientific Chemicals
Nickel Iron Molybdenum foil, 0.102mm (0.004in) thick, annealed, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel Iron Molybdenum foil, 0.102mm (0.004in) thick, annealed, Thermo Scientific Chemicals

Form: approx. 40.1g/150 x 300mm
Have Questions?
Change viewbuttonViewtableView
Quantity:
300 x 300 mm
150 x 300 mm
300 x 600 mm
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045092.HB
also known as 045092-HB
Price (USD)/ Each
84.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
99.20 
Save 14.55 (15%)
-
Add to cart
Quantity:
300 x 300 mm
Request bulk or custom format
Price (USD)/ Each
84.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
99.20 
Save 14.55 (15%)
Add to cart
Nickel Iron Molybdenum foil, 0.102mm (0.004in) thick, annealed, Thermo Scientific Chemicals
Catalog number045092.HB
Price (USD)/ Each
84.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
99.20 
Save 14.55 (15%)
-
Add to cart
Specifications
Chemical Name or MaterialNickel Iron Molybdenum foil
Name Note0.102mm (0.004 in.) thick, Annealed
Health Hazard 1H317-H350-H372
Health Hazard 2GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
View more
It is most commonly used as a high temperature, corrosion resistant metal alloy in steel, cast iron and super alloys for the military and defense industry, semiconductor industry and specialty machine shops.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only