Antimony shot, 6.35mm (0.25in) & down, 99% (metals basis), Thermo Scientific Chemicals
Antimony shot, 6.35mm (0.25in) & down, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Antimony shot, 6.35mm (0.25in) & down, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Catalog number 045095.22
also known as 045095-22
Price (USD)/ Each
40.60
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
40.60
Add to cart
Antimony shot, 6.35mm (0.25in) & down, 99% (metals basis), Thermo Scientific Chemicals
Catalog number045095.22
Price (USD)/ Each
40.60
-
Add to cart
Specifications
Chemical Name or MaterialAntimony
ColorSilver to Gray
CAS7440-36-0
AppearanceSilver Gray shot
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only