Tin shot, 3.175mm (0.125in) & down, 99% (metals basis), Thermo Scientific Chemicals
Tin shot, 3.175mm (0.125in) & down, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin shot, 3.175mm (0.125in) & down, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
Catalog number 045096.A1
also known as 045096-A1
Price (USD)/ Each
180.65
Online exclusive
201.00 
Save 20.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
180.65
Online exclusive
201.00 
Save 20.35 (10%)
Add to cart
Tin shot, 3.175mm (0.125in) & down, 99% (metals basis), Thermo Scientific Chemicals
Catalog number045096.A1
Price (USD)/ Each
180.65
Online exclusive
201.00 
Save 20.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only