Nickel gauze, 60 mesh woven from 0.18mm (0.007in) wire, Thermo Scientific Chemicals
Nickel gauze, 60 mesh woven from 0.18mm (0.007in) wire, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel gauze, 60 mesh woven from 0.18mm (0.007in) wire, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
300 x 300 mm
75 x 75 mm
150 x 150 mm
Catalog number 045099.HB
also known as 045099-HB
Price (USD)/ Each
313.65
Online exclusive
348.00 
Save 34.35 (10%)
-
Add to cart
Quantity:
300 x 300 mm
Request bulk or custom format
Price (USD)/ Each
313.65
Online exclusive
348.00 
Save 34.35 (10%)
Add to cart
Nickel gauze, 60 mesh woven from 0.18mm (0.007in) wire, Thermo Scientific Chemicals
Catalog number045099.HB
Price (USD)/ Each
313.65
Online exclusive
348.00 
Save 34.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialNickel
ColorSilver to Gray
CAS7440-02-0
Health Hazard 1H317-H351-H372
Health Hazard 3P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only