Thermo Scientific Chemicals

Scandium arc-cast pellet, 15.9mm (0.63in) dia, Thermo Scientific Chemicals

Catalog number: 045118.KF
1 Pc., Each
Thermo Scientific Chemicals

Scandium arc-cast pellet, 15.9mm (0.63in) dia, Thermo Scientific Chemicals

Catalog number: 045118.KF
1 Pc., Each
Quantity
Catalog number: 045118.KF
also known as 045118-KF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-20-2
IUPAC Name
scandium
Molecular Formula
Sc
InChI Key
SIXSYDAISGFNSX-UHFFFAOYSA-N
SMILES
[Sc]
Total Rare Earth Oxide Impurities
0.001% max.

Description

Scandium arc-cast pellet is used in aerospace industry, oil refineries as a tracing agent, and metal halide lamps/light bulbs. It is mainly used for research purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text