This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045119.KF
Price (USD)/ Each
-
Quantity:
1 Pc.
Request bulk or custom format
Specifications
Chemical Name or MaterialMagnesium cube
ColorSilver
Name NoteNominally 19mm (0.75in) each side
CAS7439-95-4
Health Hazard 1H228-H252-H261
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.