Dimethyl acetylenedicarboxylate, 95%, Thermo Scientific Chemicals
Dimethyl acetylenedicarboxylate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl acetylenedicarboxylate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 045131.36
also known as 045131-36
Price (USD)/ Each
1,002.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
1,002.00
Add to cart
Dimethyl acetylenedicarboxylate, 95%, Thermo Scientific Chemicals
Catalog number045131.36
Price (USD)/ Each
1,002.00
-
Add to cart
Chemical Identifiers
CAS762-42-5
IUPAC Name1,4-dimethyl but-2-ynedioate
Molecular FormulaC6H6O4
InChI KeyVHILMKFSCRWWIJ-UHFFFAOYSA-N
SMILESCOC(=O)C#CC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
FormLiquid
Assay (GC)> 94.0%
Dimethyl acetylenedicarboxylate as a versatile dienophile used in Diels-Alder reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only