Copper gauze, 100 mesh woven from 0.11mm (0.0045in) dia wire, Thermo Scientific Chemicals
Copper gauze, 100 mesh woven from 0.11mm (0.0045in) dia wire, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper gauze, 100 mesh woven from 0.11mm (0.0045in) dia wire, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
300 x 300 mm
150 x 150 mm
Catalog number 045186.HB
also known as 045186-HB
Price (USD)/ Each
44.65
Online exclusive
49.40 
Save 4.75 (10%)
-
Add to cart
Quantity:
300 x 300 mm
Request bulk or custom format
Price (USD)/ Each
44.65
Online exclusive
49.40 
Save 4.75 (10%)
Add to cart
Copper gauze, 100 mesh woven from 0.11mm (0.0045in) dia wire, Thermo Scientific Chemicals
Catalog number045186.HB
Price (USD)/ Each
44.65
Online exclusive
49.40 
Save 4.75 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Transition TemperatureNo Specifications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only