Thermo Scientific Chemicals
Copper plate, Oxygen-Free High Conductivity (OFHC), alloy 101, 6.35mm (0.25in) thick, Thermo Scientific Chemicals
Catalog number: 045190.GT
150 x 150 mm, Each
Thermo Scientific Chemicals
Copper plate, Oxygen-Free High Conductivity (OFHC), alloy 101, 6.35mm (0.25in) thick, Thermo Scientific Chemicals
Catalog number: 045190.GT
150 x 150 mm, Each
Quantity
Catalog number: 045190.GT
also known as 045190-GT
Price (USD)
Quantity
-
Have Questions?
Description
Copper foil, Oxygen-Free High Conductivity (OFHC), alloy 101 is used for automotive rectifiers, transistor component bases, high resistance-ratio cryogenic shunts, wave guides, hollow conductors, anodes for vacuum tubes, coaxial cable, coaxial tube, klystrons, microwave tubes, bus bars, lead-in wire, vacuum seals and glass-to-metal seals.