Indium, plasma standard solution, Specpure™ In 10μg/mL, Thermo Scientific Chemicals
Indium, plasma standard solution, Specpure™ In 10μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indium, plasma standard solution, Specpure™ In 10μg/mL, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Catalog number 045256.AE
also known as 045256-AE
Price (USD)/ Each
70.20
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
70.20
Add to cart
Indium, plasma standard solution, Specpure™ In 10μg/mL, Thermo Scientific Chemicals
Catalog number045256.AE
Price (USD)/ Each
70.20
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialIndium, plasma standard solution
ConcentrationIn in 2% HNO3
Name NoteIn 10μg/mL
Health Hazard 1H290-H315-H319
Quantity100 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only