Thermo Scientific Chemicals

Iron 57, 57Fe, plasma standard solution, Specpure™, 57Fe 10μg/mL, Thermo Scientific Chemicals

Catalog number: 045277.AD
50 mL, Each
Thermo Scientific Chemicals

Iron 57, 57Fe, plasma standard solution, Specpure™, 57Fe 10μg/mL, Thermo Scientific Chemicals

Catalog number: 045277.AD
50 mL, Each
Quantity
Catalog number: 045277.AD
also known as 045277-AD
Price (USD)
Quantity
-
Have Questions?
Assay from Supplier's CofA
⁵⁷Fe 10 (?2%)?g/ml in 2% HNO₃.
Assay from Supplier's CofA
This is a non-radioactive, "stable" isotope.
Form
Liquid
Assay from Supplier's CofA
Further Ingredients: 2% HNO₃ CAS 7697-37-2, EEC No 231-714-2

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text