Chemical Name or MaterialLead, NIST Pb, plasma standard solution
Name NoteNIST Pb 100μg/mL
Health Hazard 1H290-H315-H319
Quantity50 mL
Health Hazard 2GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
View more