Specifications
Chemical Name or MaterialThallium 203, 203Tl, plasma standard solution
Name Note203Tl 10μg/mL
Health Hazard 1H290-H314-H335
Quantity50 mL
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
View more