Sulfur in Light Mineral Oil standard solution, Specpure™, 5μg/g (0.0005%), Thermo Scientific Chemicals
Sulfur in Light Mineral Oil standard solution, Specpure™, 5μg/g (0.0005%), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfur in Light Mineral Oil standard solution, Specpure™, 5μg/g (0.0005%), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Catalog number 045315.AE
also known as 045315-AE
Price (USD)/ Each
114.00
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
114.00
Add to cart
Sulfur in Light Mineral Oil standard solution, Specpure™, 5μg/g (0.0005%), Thermo Scientific Chemicals
Catalog number045315.AE
Price (USD)/ Each
114.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialSulfur in Light Mineral Oil standard solution
Name Note5μg/g (0.0005%)
Health Hazard 1Danger
Quantity100 mL
Health Hazard 2GHS H Statement
H350
May cause cancer.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only