Chemical Name or MaterialSulfur in Isooctane standard solution
Name Note750μg/g (0.0750%)
Health Hazard 1H225-H304-H315-H319-H336
Quantity100 mL
Health Hazard 2GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
View more