Lead in Isooctane standard solution, Specpure™, 0μg/g(0.000g/gal), Thermo Scientific Chemicals
Lead in Isooctane standard solution, Specpure™, 0μg/g(0.000g/gal), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead in Isooctane standard solution, Specpure™, 0μg/g(0.000g/gal), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 g
Catalog number 045330.22
also known as 045330-22
Price (USD)/ Each
125.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
125.00
Add to cart
Lead in Isooctane standard solution, Specpure™, 0μg/g(0.000g/gal), Thermo Scientific Chemicals
Catalog number045330.22
Price (USD)/ Each
125.00
-
Add to cart
Chemical Identifiers
CAS540-84-1
IUPAC Name2,2,4-trimethylpentane
Molecular FormulaC8H18
InChI KeyNHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILESCC(C)CC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
CommentFor ASTM D5059 Pt. A and Pt. C.
CommentBlank
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only