Osmium(VIII) oxide, 2% aq. soln., Thermo Scientific Chemicals
Osmium(VIII) oxide, 2% aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Osmium(VIII) oxide, 2% aq. soln., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
2 mL
Catalog number 045384.AB
also known as 045384-AB
Price (USD)/ Each
114.65
Online exclusive
127.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
10 mL
Request bulk or custom format
Price (USD)/ Each
114.65
Online exclusive
127.00 
Save 12.35 (10%)
Add to cart
Osmium(VIII) oxide, 2% aq. soln., Thermo Scientific Chemicals
Catalog number045384.AB
Price (USD)/ Each
114.65
Online exclusive
127.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS20816-12-0
IUPAC Nametetraoxoosmium
Molecular FormulaO4Os
InChI KeyVUVGYHUDAICLFK-UHFFFAOYSA-N
SMILESO=[Os](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Pale yellow
Assay from Supplier's CofA1.4-1.6% Os (1.9-2.1% as OsO₄) in water
Osmium(VIII) oxide is widely used as an oxidant in organic synthesis to oxidize alkenes to vicinal diols as well as in the sharpless oxyamination to give vicinal amino-alcohols. It is involved in the oxidative cleavage of alkenes (Lemieux-Johnson oxidation) in association with sodium periodate. It finds application in transmission electron microscopy (TEM) as a staining agent. It is involved in the promotion of oxidative cleavage of graphene layers in carbon nanotubes and as an intermediate in osmium ore refining. As a lipid stain, it is useful in scanning electron microscopy (SEM) as an alternative to sputter coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only