Thermo Scientific Chemicals

Osmium(VIII) oxide, 4% aq. soln., Thermo Scientific Chemicals

Catalog number: 045385.9W
2 mL, Each
Thermo Scientific Chemicals

Osmium(VIII) oxide, 4% aq. soln., Thermo Scientific Chemicals

Catalog number: 045385.9W
2 mL, Each
Quantity
Catalog number: 045385.9W
also known as 045385-9W
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20816-12-0
IUPAC Name
tetraoxoosmium
Molecular Formula
O4Os
InChI Key
VUVGYHUDAICLFK-UHFFFAOYSA-N
SMILES
O=[Os](=O)(=O)=O
Appearance (Color)
Pale yellow
Assay (special)
3.0 ?0.1%
Form
Liquid

Description

Osmium(VIII) oxide is widely used as an oxidant in organic synthesis to oxidize alkenes to vicinal diols as well as in the sharpless oxyamination to give vicinal amino-alcohols. It is involved in the oxidative cleavage of alkenes (Lemieux-Johnson oxidation) in association with sodium periodate. It finds application in transmission electron microscopy (TEM) as a staining agent. It is involved in the promotion of oxidative cleavage of graphene layers in carbon nanotubes and as an intermediate in osmium ore refining. As a lipid stain, it is useful in scanning electron microscopy (SEM) as an alternative to sputter coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text