D-Glucose dehydrogenase
D-Glucose dehydrogenase
Thermo Scientific Chemicals

D-Glucose dehydrogenase

 
Have Questions?
Quantity:
1 kU
Catalog number 045402.4I
also known as 045402-4I
Price (USD)/ Each
130.65
Online exclusive
145.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
1 kU
Request bulk or custom format
Price (USD)/ Each
130.65
Online exclusive
145.00 
Save 14.35 (10%)
Add to cart
D-Glucose dehydrogenase
Catalog number045402.4I
Price (USD)/ Each
130.65
Online exclusive
145.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9028-53-9
SpecificationsSpecification SheetSpecification Sheet
Comment1 unit will reduce 1.0µmol of NAD to NADH per minute at pH 7, 25°C
Appearance (Color)White to pale cream
FormPowder
Assay from Supplier's CofAActivity: ≥U/mg
Diagnostics and cofactor regeneration of NAD(P)H in reductive processes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only