Silver cyanate, 98%, Thermo Scientific Chemicals
Silver cyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver cyanate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 045411.06
also known as 045411-06
Price (USD)/ Each
74.20
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
74.20
Add to cart
Silver cyanate, 98%, Thermo Scientific Chemicals
Catalog number045411.06
Price (USD)/ Each
74.20
-
Add to cart
Chemical Identifiers
CAS3315-16-0
IUPAC Namesilver(1+) cyanate
Molecular FormulaCAgNO
InChI KeyDWBPIWPCOSHWCK-UHFFFAOYSA-M
SMILES[Ag+].[O-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.6% min
Silver cyanate is a chemosterilant in adult house flies. It is also used as a reagent to prepare 1-(3-thianyl)uracil and 9-(3-thianyl)adenine derivatives, which has potential anti-human immunodeficiency virus (HIV) and antitumour activity.It is used as a silver plating solution in the electroplating process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only