Thermo Scientific™

Silver cyanate, 98%, Thermo Scientific Chemicals

Catalog number: 045411.14
25 g, Each
Thermo Scientific™

Silver cyanate, 98%, Thermo Scientific Chemicals

Catalog number: 045411.14
25 g, Each
Quantity
Catalog number: 045411.14
also known as 045411-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3315-16-0
IUPAC Name
silver(1+) cyanate
Molecular Formula
CAgNO
InChI Key
DWBPIWPCOSHWCK-UHFFFAOYSA-M
SMILES
[Ag+].[O-]C#N
Assay (unspecified)
97.6% min

Description

Silver cyanate is a chemosterilant in adult house flies. It is also used as a reagent to prepare 1-(3-thianyl)uracil and 9-(3-thianyl)adenine derivatives, which has potential anti-human immunodeficiency virus (HIV) and antitumour activity.It is used as a silver plating solution in the electroplating process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text