Silver sulfide, 99.999% (metals basis), Thermo Scientific Chemicals
Silver sulfide, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver sulfide, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 045458.14
also known as 045458-14
Price (USD)/ Each
480.65
Online exclusive
534.00 
Save 53.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
480.65
Online exclusive
534.00 
Save 53.35 (10%)
Add to cart
Silver sulfide, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number045458.14
Price (USD)/ Each
480.65
Online exclusive
534.00 
Save 53.35 (10%)
-
Add to cart
Chemical Identifiers
CAS21548-73-2
IUPAC Namedisilver(1+) sulfanediide
Molecular FormulaAg2S
InChI KeyXUARKZBEFFVFRG-UHFFFAOYSA-N
SMILES[S--].[Ag+].[Ag+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Silver sulfide is used as photosensitizer in photography, anti-bacterial and anti-microbial agents. It is also useful as qualitative inorganic analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only