Thermo Scientific Chemicals

Zinc phosphide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 045459.04
2 g, Each
Thermo Scientific Chemicals

Zinc phosphide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 045459.04
2 g, Each
Quantity
Catalog number: 045459.04
also known as 045459-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-84-7
IUPAC Name
zinc phosphide
Molecular Formula
H2P2Zn3
InChI Key
HOKBIQDJCNTWST-UHFFFAOYSA-N
SMILES
[Zn].P=[Zn].P=[Zn]
Particle Size
6mm and down
Appearance (Color)
Gray
Total Metal Impurities
≤0.001%
Form
Pieces

Description

Zinc phosphide has useful applications in photovoltaic cells. It is also employed as a laboratory chemical. It plays an important role in medicine and pharmaceuticals. It acts as a fumigant in grain storage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text