Thermo Scientific Chemicals

Lead(II) sulfide, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 045462.09
10 g, Each
Thermo Scientific Chemicals

Lead(II) sulfide, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 045462.09
10 g, Each
Quantity
Catalog number: 045462.09
also known as 045462-09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1314-87-0
IUPAC Name
λ²-lead(2+) sulfanediide
Molecular Formula
PbS
InChI Key
FILXVLQXZYXBSJ-UHFFFAOYSA-N
SMILES
[S--].[Pb++]
Total Metal Impurities
0.005% max.

Description

Lead(II) sulfide is used in infrared detectors, photo optic applications, slip property modifier used in friction industry for enhancing heat conduction and regulating friction coefficient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text