Thermo Scientific Chemicals

Tin(IV) isopropoxide, 99% (metals basis), 10% w/v in isopropanol, Thermo Scientific Chemicals

Catalog number: 045480.AA
5 mL, Each
Thermo Scientific Chemicals

Tin(IV) isopropoxide, 99% (metals basis), 10% w/v in isopropanol, Thermo Scientific Chemicals

Catalog number: 045480.AA
5 mL, Each
Quantity
Catalog number: 045480.AA
also known as 045480-AA
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1184-61-8
IUPAC Name
tetrakis(propan-2-ol) tin
Molecular Formula
C12H32O4Sn
InChI Key
VUDAJANAMZGVOP-UHFFFAOYSA-N
SMILES
[Sn].CC(C)O.CC(C)O.CC(C)O.CC(C)O
Appearance (Color)
Yellow to red
Form
Liquid
Concentration (w/v)
9.5-10.5%
Total Metal Impurities
≤1%

Description

Tin(IV) isopropoxide is used as a precursor for the preparation of tin oxide nanoparticles, thin films, indium tin oxide (ITO) films , which shows desired electrochemical and conducting properties. Doped tin oxide nanoparticles and thin films find application as gas sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text