Thermo Scientific Chemicals

Methyl cellulose, viscosity 15 cPs, Thermo Scientific Chemicals

Catalog number: 045490.A4
2.5 kg, Each
Thermo Scientific Chemicals

Methyl cellulose, viscosity 15 cPs, Thermo Scientific Chemicals

Catalog number: 045490.A4
2.5 kg, Each
Quantity
Catalog number: 045490.A4
also known as 045490-A4
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9004-67-5
IUPAC Name
3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
Molecular Formula
C20H38O11
InChI Key
YLGXILFCIXHCMC-UHFFFAOYNA-N
SMILES
COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC
Form
Powder
Appearance (Color)
White to pale yellow
Viscosity
12-18 cPs for a 2% aqueous solution at 20?C

Description

Methyl cellulose is used as an emulsifying and suspending agent in cosmetics, pharmaceutics and the chemical industry. It is used therapeutically as a bulk laxative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text