Thermo Scientific Chemicals

Silver(I) behenate, Thermo Scientific Chemicals

Catalog number: 045494.06
5 g, Each
Thermo Scientific Chemicals

Silver(I) behenate, Thermo Scientific Chemicals

Catalog number: 045494.06
5 g, Each
Quantity
Catalog number: 045494.06
also known as 045494-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2489-05-6
IUPAC Name
silver(1+) docosanoate
Molecular Formula
C22H43AgO2
InChI Key
AQRYNYUOKMNDDV-UHFFFAOYSA-M
SMILES
[Ag+].CCCCCCCCCCCCCCCCCCCCCC([O-])=O
Assay (Argentometric Titration)
≥95.0 to ≤105.0%
Appearance (Color)
White to green to orange
Form
Crystals or powder or crystalline powder

Description

Silver(I) behenate is used as a calibration standard in X-ray spectrophotometer to an absolute X-ray energy scale. It is utilized for low-angle diffraction standard in X-ray crystallography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text