Thermo Scientific Chemicals

Silver(I) chromate, 96%, Thermo Scientific Chemicals

Catalog number: 045495.06
5 g, Each
Thermo Scientific Chemicals

Silver(I) chromate, 96%, Thermo Scientific Chemicals

Catalog number: 045495.06
5 g, Each
Quantity
Catalog number: 045495.06
also known as 045495-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7784-01-2
IUPAC Name
disilver(1+) dioxochromiumbis(olate)
Molecular Formula
Ag2CrO4
InChI Key
OJKANDGLELGDHV-UHFFFAOYSA-N
SMILES
[Ag+].[Ag+].[O-][Cr]([O-])(=O)=O
Form
Powder
Assay from Suppliers CofA
≥95.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text