Thermo Scientific Chemicals

Silver(I) citrate hydrate, Thermo Scientific Chemicals

Catalog number: 045496.06
5 g, Each
Thermo Scientific Chemicals

Silver(I) citrate hydrate, Thermo Scientific Chemicals

Catalog number: 045496.06
5 g, Each
Quantity
Catalog number: 045496.06
also known as 045496-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
206986-90-5
IUPAC Name
trisilver(1+) 2-hydroxypropane-1,2,3-tricarboxylate
Molecular Formula
C6H5Ag3O7
InChI Key
QUTYHQJYVDNJJA-UHFFFAOYSA-K
SMILES
[Ag+].[Ag+].[Ag+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Appearance (Color)
White to gray
Form
Powder
Assay (Argentometric Titration)
≥94.0 to ≤103.0% (Dry wt. basis)
Water Content (Karl Fischer Titration)
≤6.0%

Description

Silver(I) citrate hydrate can be very attractive for the future biomedical or pharmaceutical/therapeutic applications. It can be used as an antimicrobial agent and enhancers of antioxidants and anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text