Silver(I) citrate hydrate, Thermo Scientific Chemicals
Silver(I) citrate hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver(I) citrate hydrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 045496.22
also known as 045496-22
Price (USD)/ Each
783.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
783.00
Add to cart
Silver(I) citrate hydrate, Thermo Scientific Chemicals
Catalog number045496.22
Price (USD)/ Each
783.00
-
Add to cart
Chemical Identifiers
CAS206986-90-5
IUPAC Nametrisilver(1+) 2-hydroxypropane-1,2,3-tricarboxylate
Molecular FormulaC6H5Ag3O7
InChI KeyQUTYHQJYVDNJJA-UHFFFAOYSA-K
SMILES[Ag+].[Ag+].[Ag+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to gray
FormPowder
Assay (Argentometric Titration)≥94.0 to ≤103.0% (Dry wt. basis)
Water Content (Karl Fischer Titration)≤6.0%
Silver(I) citrate hydrate can be very attractive for the future biomedical or pharmaceutical/therapeutic applications. It can be used as an antimicrobial agent and enhancers of antioxidants and anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only