Thermo Scientific Chemicals

Nickel sulfide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 045498.06
5 g, Each
Thermo Scientific Chemicals

Nickel sulfide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 045498.06
5 g, Each
Quantity
Catalog number: 045498.06
also known as 045498-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12035-51-7
IUPAC Name
disulfanylidenenickel
Molecular Formula
NiS2
InChI Key
HNDMNBUITRNJKK-UHFFFAOYSA-N
SMILES
S=[Ni]=S
Total Metal Impurities
1% max.

Description

Nickel sulfide is used as a semiconductor material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text