Thermo Scientific Chemicals

Bismuth 2-ethylhexanoate acid, Thermo Scientific Chemicals

Catalog number: 045500.14
25 g, Each
Thermo Scientific Chemicals

Bismuth 2-ethylhexanoate acid, Thermo Scientific Chemicals

Catalog number: 045500.14
25 g, Each
Quantity
Catalog number: 045500.14
also known as 045500-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67874-71-9
IUPAC Name
bismuth(3+) tris(2-ethylhexanoate)
Molecular Formula
C24H45BiO6
InChI Key
NUMHJBONQMZPBW-UHFFFAOYNA-K
SMILES
[Bi+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Appearance (Color)
Pale yellow to brown
Assay from Supplier's CofA
≥60.0%
Form
Liquid to viscous liquid

Description

Bismuth 2-ethylhexanoate is used as an industrial lubricant, dessicant, and polyurethane catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text