Thermo Scientific™

Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II), Pd 15%, Thermo Scientific™

Catalog number: 045511.03
1 g, Each
Thermo Scientific™

Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II), Pd 15%, Thermo Scientific™

Catalog number: 045511.03
1 g, Each
Quantity
Catalog number: 045511.03
also known as 045511-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
887919-35-9
IUPAC Name
bis(4-(di-tert-butylphosphanyl)-N,N-dimethylaniline); dichloropalladium
Molecular Formula
C32H56Cl2N2P2Pd
InChI Key
DWOZNANUEDYIOF-UHFFFAOYSA-L
SMILES
Cl[Pd]Cl.CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C.CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C
Proton NMR
Pass
Phosphorus-31 NMR
Pass
Assay (unspecified)
C 53.30-55.30%
Assay (unspecified)
H 7.50-8.50%
Assay (unspecified)
Pd 14.00-16.00%

Description

Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) is a highly active, air-stable catalyst for Suzuki-Miyaura cross-coupling with aryl halides including 5- and 6-membered heteroaryl chlorides.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text