Thermo Scientific™

Palladium(II) nitrate, solution, Pd 4-5% w/w (cont. Pd), Thermo Scientific Chemicals

Catalog number: 045512.NB
5 g, Each
Thermo Scientific™

Palladium(II) nitrate, solution, Pd 4-5% w/w (cont. Pd), Thermo Scientific Chemicals

Catalog number: 045512.NB
5 g, Each
Quantity
Catalog number: 045512.NB
also known as 045512-NB
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10102-05-3
IUPAC Name
palladium(2+) dinitrate
Molecular Formula
N2O6Pd
InChI Key
GPNDARIEYHPYAY-UHFFFAOYSA-N
SMILES
[Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
Pale yellow to red-brown to brown
Assay (unspecified)
4.0-5.2% Pd Content
Form
Liquid

Description

Palladium(II) nitrate dehydrate is used as a reactant for doping activated carbon for catalysis. It acts as a catalyst for hydrogenation of 1-heptyne and naphthalene. It serves as an important raw material for the preparation of platinum-palladium carbon alloy nanocatalysts for methanol-tolerant oxygen reduction reaction in fuel cells and in the synthesis of copper palladium alloy thin films. Furthermore, it is used in the synthesis of di-phenyl sulfide-modified palladium-titanium dioxide catalysts for acetylene hydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text