Magnesium silicide, 99.99% (metals basis), Thermo Scientific Chemicals
Magnesium silicide, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium silicide, 99.99% (metals basis), Thermo Scientific Chemicals

Air and Moisture Sensitive; Decomposes in water, HCl
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number 045518.30
also known as 045518-30
Price (USD)/ Each
1,085.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
1,085.00
Add to cart
Magnesium silicide, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number045518.30
Price (USD)/ Each
1,085.00
-
Add to cart
Chemical Identifiers
CAS22831-39-6
IUPAC Namedimagnesiosilane
Molecular FormulaMg2Si
InChI KeyYTHCQFKNFVSQBC-UHFFFAOYSA-N
SMILES[Mg]=[Si]=[Mg]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Magnesium silicide is used to produce alloys such as aluminum alloys. It is also used in food, beverages, as a flavor enhancer and as an intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only