Thermo Scientific Chemicals

Devarda's Alloy, granular

Catalog number: 045526.22
100 g, Each
Thermo Scientific Chemicals

Devarda's Alloy, granular

Catalog number: 045526.22
100 g, Each
Quantity
Catalog number: 045526.22
also known as 045526-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
8049-11-4
IUPAC Name
nonaaluminium decacopper zinc
Molecular Formula
Al9Cu10Zn
InChI Key
MPKKOQSNGWAMJD-UHFFFAOYSA-N
SMILES
[Al].[Al].[Al].[Al].[Al].[Al].[Al].[Al].[Al].[Cu].[Cu].[Cu].[Cu].[Cu].[Cu].[Cu].[Cu].[Cu].[Cu].[Zn]
Assay (unspecified)
44-46% Al
Assay (unspecified)
4-6% Zn
Assay (unspecified)
49-51% Cu

Description

Devarda's Alloy is used as reducing agent in analytical chemistry for the determination of nitrates after their reduction to ammonia under alkaline conditions. It is also used in the quantitative or qualitative analysis of nitrates in agriculture and soil science before the development of ion chromatography. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text